Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-NK1.1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: PK136]

Supplier: Southern Biotechnology
1805-30
103259-962EA 300.77 USD InStock
103259-962
Anti-NK1.1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: PK136]
Antibodies
NK1.1, a member of the NKR-P1 family of cell surface receptors, is a type II integral membrane glycoprotein with a C-type lectin domain. It is expressed as a disulfide-linked homodimer on all NK cells as well as subsets of thymocytes and peripheral T lymphocytes in selected strains of mice (e.g., C57BL/6, NZB, and CE). NK1.1 mediates cellular activation and differentiation, and is thought to have a particular role in generating Th2 cells. This product does not react with NK cells of BALB/c mice.

Type: Primary
Antigen: NK1.1
Clonality: Monoclonal
Clone: PK136
Conjugation: Alexa Fluor® 488
More Product Information
Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty