Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD61 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PM6/13]

Supplier: Southern Biotechnology
9470-09
103265-276EA 311.23 USD InStock
103265-276
Anti-CD61 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PM6/13]
Antibodies
CD61 is the integrin β3 subunit, a 110 kDa transmembrane glycoprotein expressed on platelets, megakaryocytes, and endothelial cells. Integrin β3 associates with gpIIb (CD41) to form the CD41/CD61 complex that mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the αVβ3 vitronectin receptor.

Type: Primary
Antigen: CD61
Clonality: Monoclonal
Clone: PM6/13
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty