Anti-Cytokeratin 19 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB39g]
Supplier: Southern Biotechnology
10090-05
103265-476EA 453.77 USD InStock
103265-476
Anti-Cytokeratin 19 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SB39g]
Antibodies
Keratins are a family of intermediate filament proteins that assemble into filaments through forming heterodimers of one type I keratin (keratins 9 to 23) and one type II keratin (keratins 1-8). The two keratin types share only 30% sequence homology. Keratins demonstrate tissue- and differentiation-specific expression profiles. Cytokeratin 19 is the smallest human keratin. It is found in many simple and some non-keratinizing stratified squamous epithelia and is often co-expressed with keratin 7. Cytokeratin 19 has been implicated as a marker for stem cells in some tissues such as hairMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty