Glutathione Cell-Based Detection Kit (Blue Fluorescence), Cayman Chemical Company
Supplier: CAYMAN CHEMICAL COMPANY, INC.
600360-96
75816-984EA 388.1 USD InStock
75816-984
Glutathione Cell-Based Detection Kit (Blue Fluorescence), Cayman Chemical Company
Assays Cellular Assays
For rapid and easy determination of GSH in whole cells.
  • Utilizes MCB, a highly fluorescent GSH probe
  • Includes a standard curve for accurate quantification of GSH from cell lysates
  • Rapid assay; get results in 2 hours
Glutathione (GSH) is a low molecular-weight tripeptide thiol that serves as a primary cellular anti-oxidant and plays a fundamental role in the elimination of environmental toxins. During early apoptosis, cells may exclude GSH, causing a decrease in intracellular GSH levels. The intracellular level of GSH is thus used as an indicator of cell health. Cayman’s Cell-Based Glutathione Assay Kit (Blue Fluorescence) employs a cell-permeable dye, monochlorobimane (MCB), which reacts with GSH to generate a highly fluorescent product that can be measured using excitation and emission wavelengths of 380 and 480 nm, respectively. The kit can be easily adapted to screening programs for therapeutic compounds regulating intracellular GSH levels.
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Collapse All
Contents Supplier No. VWR Catalog Number Unit Price Quantity
Kit contains: Monochlorobimane, Standard, Buffer, and Plate 600360-96 75816-984 Each
Retrieving
Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty