Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD5 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 4H8E5]

Supplier: Southern Biotechnology
1546-09 1546-09L
100243-160EA 241.92 USD InStock
100243-160 100243-162
Anti-CD5 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 4H8E5]
Antibodies
CD5, also known as Lyt-1, is a monomeric type I transmembrane glycoprotein expressed on thymocytes, T lymphocytes, and a subset of B lymphocytes, but not on natural killer (NK) cells. It has been identified as the major ligand of the B-cell antigen CD72. The frequency of CD5+ B cells exhibits strain-dependent variation, and the phenotypic, anatomical, functional, developmental, and pathological characteristics of the CD5+ B cells suggest that they may represent a distinct lineage, known as B-1 cells. Binding of CD5 on the T cell surface can augment alloantigen- or mitogen-induced lymphocyte proliferation and induces increased cytosolic free calcium, IL-2 secretion, and IL-2R expression. It has been proposed that CD5 negatively regulates signal transduction mediated by the T-cell and B-cell receptors.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4H8E5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty