Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD11b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 3A33]

Supplier: Southern Biotechnology
1560-09 1560-09L
100243-212EA 211.85 USD InStock
100243-212 100243-214
Anti-CD11b Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 3A33]
Antibodies
CD11b (integrin αM subunit) combines with CD18 (integrin β2 subunit) to form the integrin Mac-1, also known as complement receptor 3 (CR3), which mediates adhesion to C3bi and ICAM-1 (CD54). Mac-1 is expressed at varying levels on granulocytes, macrophages, dendritic cells, NK cells, and B-1 cells in the peritoneal and pleural cavities. In addition to its role in binding C3bi on opsonized targets and mediation of the subsequent ingestion process, Mac-1 is important as an adhesion molecule in the transendothelial migration of monocytes and neutrophils.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: 3A33
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty