Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD23 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2G8]

Supplier: Southern Biotechnology
1585-09 1585-09L
100243-316EA 272 USD InStock
100243-316 100243-318
Anti-CD23 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2G8]
Antibodies
The CD23 antigen is the low affinity IgE Fc receptor, which is a 45-49 kDa protein with 38 and 28 kDa fragments. It is expressed on most mature, conventional B cells (but not on peritoneal CD5+ B cells) and can also be found on the surface of T cells, macrophages and platelets. It is distinct from the high affinity IgE receptors found on basophils and mast cells which mediate allergic reactions. The low affinity receptors are thought to play a role in isotype specific immunoregulation. The regulation of CD23 surface expression appears to be integral with the complex IgE system which involves interactions of cells, cytokines, antibodies and regulatory factors. CD23 has been described as a “membrane bound cytokine," in that the soluble cleavage products of CD23 are themselves able to act as cytokines in vitro.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 2G8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty