Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD31 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 390]

Supplier: Southern Biotechnology
1625-09 1625-09L
100243-420EA 287.69 USD InStock
100243-420 100243-422
Anti-CD31 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 390]
Antibodies
CD31, also known as platelet endothelial cell adhesion molecule-1 (PECAM-1), is a type I integral membrane glycoprotein and a member of the immunoglobulin superfamily of cell surface receptors. It is constitutively expressed on the surface of endothelial cells and concentrated at the junction between them. It is also weakly expressed on many peripheral lymphoid cells and platelets. CD31 interacts homotypically in cell adhesion assays. The monoclonal antibody 390 inhibits the aggregation of L cells transfected with a variant form of CD31.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 390
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty