Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD45R Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RA3-6B2]

Supplier: Southern Biotechnology
1665-02 1665-02S
100243-524EA 277.23 USD InStock
100243-524 100243-526
Anti-CD45R Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RA3-6B2]
Antibodies
CD45R, also known as B220, a member of the protein tyrosine phosphate family and a major cell surface glycoprotein, represents a restricted form of the CD45 family which primarily recognizes only cells of B-lineage from pro-B cell through mature B lymphocytes and prior to the availability of anti-CD19 monoclonal antibodies was commonly used as a pan B-cell marker. It also reacts with certain activated T cells as well as non-MHC-restricted lytically active lymphokine-activated killer (LAK) cells. In vivo administration of RA3-6B2 has been shown to affect differentiation of both T and B cells in normal mice and reduce the level of anti-DNA antibodies and lymphadenopathies in MRL/lpr mice.

Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human, Feline

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty