Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD45RB Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C363.16A]

Supplier: Southern Biotechnology
1675-02S 1675-02
100243-550EA 129.46 USD InStock
100243-550 100243-548
Anti-CD45RB Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C363.16A]
Antibodies
CD45RB represents an exon B-dependent form of CD45 glycoprotein. It is expressed at high density on peripheral B cells, the suppressor/cytotoxic subpopulation of T lymphocytes, a subset of helper T cells, most thymocytes, and at low density on macrophages and dendritic cells.

Type: Primary
Antigen: CD45RB
Clonality: Monoclonal
Clone: C363.16A
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty