Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD62L Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEL-14]

Supplier: Southern Biotechnology
1705-09 1705-09L
100243-594EA 281.15 USD InStock
100243-594 100243-596
Anti-CD62L Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEL-14]
Antibodies
CD62L, also known as L-selectin, is a type I transmembrane glycoprotein with lectin-like and epidermal growth factor-like domains. It binds to sialylated oligosaccharide determinants on high endothelial venules (HEV) in peripheral lymph nodes. In the mouse, CD62L is expressed on most thymocytes and on peripheral leukocytes, including B and T lymphocytes, neutrophils, monocytes, and eosinophils. This member of the selectin family of cell adhesion molecules appears to be required for lymphocyte homing to peripheral lymph nodes and for leukocyte extravasation at sites of inflammation. Expression of CD62L on lymphocytes and neutrophils is rapidly down-regulated upon cell activation and the level of CD62L expression, along with other markers, distinguishes naive T cells from effector/memory T cells. The monoclonal antibody MEL-14 blocks in vitro binding of lymphocytes to peripheral lymph node HEV and inhibits in vivo lymphocyte extravasation into peripheral lymph nodes and the late stages of leukocyte rolling.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: MEL-14
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty