Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD45RC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C455.1F]

Supplier: Southern Biotechnology
1820-02 1820-02S
100243-934EA 507.38 USD InStock
100243-934 100243-936
Anti-CD45RC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C455.1F]
Antibodies
CD45RC represents an exon C-dependent isoform of CD45 glycoprotein. It is expressed at high density on all B lymphocytes, at intermediate density on most CD8+ splenic T cells, and on two subpopulations of CD4+ splenic T cells with high and low density of expression. CD45RC is also present on a small subset of thymocytes.

Type: Primary
Antigen: CD45RC
Clonality: Monoclonal
Clone: C455.1F
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty