Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-Ly1]

Supplier: Southern Biotechnology
9350-02 9350-02S
100245-918EA 239.31 USD InStock
100245-918 103265-710
Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: B-Ly1]
Antibodies
CD20 is a 33-37 kDa unglycosylated phosphoprotein. Although not a member of the tetraspanin superfamily of cell surface receptors, it crosses the cell membrane four times. It is expressed on all stages of B cell differentiation except plasma cells. The CD20 molecule is involved in regulation of B cell differentiation presumably via its reported function as a Ca++ channel subunit.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: B-Ly1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Feline

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty