Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD56 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM 188]

Supplier: Southern Biotechnology
9456-02 9456-02S
100245-992EA 269.38 USD InStock
100245-992 103265-264
Anti-CD56 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM 188]
Antibodies
CD56, an isoform of the neural cell adhesion molecule (NCAM), is a heavily glycosylated type I transmembrane protein. On human hematopoietic cells its expression is restricted to NK cells and a subpopulation of T lymphocytes (known as large granular lymphocytes) which demonstrate natural killer activity. While it is clear that NCAM plays a role in homotypic adhesion of neuronal cells, the significance of CD56 expression on NK cells is not clear and remains controversial.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: MEM 188
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty