Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD11c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3.9]

Supplier: Southern Biotechnology
9551-02 9551-02S
100246-140EA 194.85 USD InStock
100246-140 103265-304
Anti-CD11c Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3.9]
Antibodies
CD11c is a 150 kDa adhesion glycoprotein that associates with β2 integrin to form the CD11c/CD18 heterodimer that binds CD54 (ICAM-1) and fibrinogen. It is expressed on NK cells, a subset of B and T cells, granulocytes, monocytes, and macrophages. It functions in cytotoxic T cell killing and in neutrophil and monocyte adhesion to endothelium.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus, Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty