Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD33 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: WM53]

Supplier: Southern Biotechnology
9590-09 9590-09S
100246-212EA 299.46 USD InStock
100246-212 103265-380
Anti-CD33 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: WM53]
Antibodies
CD33 is a 67 kDa type I transmembrane glycoprotein and a member of the sialoadhesin family of cell surface receptors. It is absent from pluripotent stem cells but appears on myelomonocytic precursors after CD34. It then continues to be expressed on both the myeloid and monocyte lineages. While it has been reported that CD33 can function as a sialic acid-dependent cell adhesion molecule, cells expressing CD33 require desialylation before they can bind cells bearing the appropriate sialoglycoconjugates. This suggests that inhibitory cis interactions may regulate or block any adhesion function.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty