Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD45 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: F10-89-4]

Supplier: Southern Biotechnology
9625-02 9625-02S
100246-272EA 190.92 USD InStock
100246-272 103265-406
Anti-CD45 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: F10-89-4]
Antibodies
CD45, also known as the leukocyte common antigen (LCA), exists as four different isoforms (180, 190, 205, 220 kDa) that are generated by alternative splicing of three exons. LCA is found on all cells of hematopoietic origin, except erythrocytes and platelets. CD45 expression is required for T cell and B cell antigen receptor-mediated activation.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: F10-89-4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Horse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty