Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD55 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 143-30]

Supplier: Southern Biotechnology
9661-09 9661-09S
100246-330EA 329.54 USD InStock
100246-330 103260-214
Anti-CD55 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 143-30]
Antibodies
CD55, also known as decay accelerating factor (DAF), is an 80 kDa GPI-anchored single chain glycoprotein that is expressed on all cells in contact with serum including all hematopoietic cells and the vascular endothelium. A soluble form of CD55 is present in plasma and body fluids. CD55 is a member of the regulator of complement activity (RCA) family of proteins and is thought to play a role in the protection of cells from damage by autologous complement by preventing the amplification steps of the complement cascade. CD55 expression increases upon T cell activation and antibodies to CD55 are mitogenic in the presence of phorbol esters.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 143-30
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty