Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD107a Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H4A3]

Supplier: Southern Biotechnology
9835-02 9835-02S
100246-516EA 291.62 USD InStock
100246-516 103265-426
Anti-CD107a Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H4A3]
Antibodies
CD107a, also known as lysosomal-associated membrane protein 1 (LAMP-1), is a heavily glycosylated type I transmembrane protein that constitutes the major sialoglycoproteins on lysosomal membranes. It is a ligand for galaptin, an S-type lectin present in extracellular matrix, through its recognition of acetyllactosamine oligosaccharide chains, and is a ligand for E-selectin-mediated cell adhesion. CD107a is expressed by activated T cells, macrophages, dendritic cells, activated platelets, tonsillar epithelium, and some tumor cell lines, including U937 and KG1a. It is also a widely expressed intracellular antigen. LAMP-1 may function in protecting the inner surface of the lysosomal membrane by forming a barrier to lysosomal hydrolases. The upregulation of both CD107a and CD107b on the surface of tumor cell lines has been associated with their enhanced metastatic potential where they may increase adhesion to extracellular matrix and endothelium.

Type: Primary
Antigen: CD107a
Clonality: Monoclonal
Clone: H4A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus, African Green Monkey, Pigtail Macaque, Sooty Mangabey

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty