Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgA Rat Antibody (RPE (R-Phycoerythrin)) [clone: 11-44-2]

Supplier: Southern Biotechnology
1165-09L 1165-09
100242-850EA 447.23 USD InStock
100242-850 100242-848
Anti-IgA Rat Antibody (RPE (R-Phycoerythrin)) [clone: 11-44-2]
Antibodies
PE Anti-Mouse IgA antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: 11-44-2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty