Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD3 epsilon Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C363.29B]

Supplier: Southern Biotechnology
1535-02S 1535-02
100243-096EA 121.62 USD InStock
100243-096 100243-094
Anti-CD3 epsilon Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C363.29B]
Antibodies
CD3ε, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chain ranging in size from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). It is expressed on all T cells of all mouse strains. CD3 plays a major role in signaling during antigen recognition, leading to T-cell activation. The monoclonal antibody C363.29B recognizes an epitope on the 25 kDa ε chain of the CD3/TCR complex. In the presence of Fc receptor-bearing accessory cells, soluble C363.29B can activate primed and naïve T cell in vitro. Immobilized C363.2B9 monoclonal antibody can also activate both normal T lymphocytes and cloned T cell lines provided the appropriate accessory signals are present. The monoclonal antibody is cytolytic, easily used for cell surface staining, and a good immunoprecipitating antibody.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: C363.29B
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty