Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD30 Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: mCD30.1]

Supplier: Southern Biotechnology
1620-09L 1620-09
100243-410EA 371.38 USD InStock
100243-410 100243-408
Anti-CD30 Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: mCD30.1]
Antibodies
CD30 is a type I transmembrane protein and a member of the tumor necrosis factor receptor superfamily of proteins. Murine CD30 is expressed predominantly in the thymus and it is inducible in mouse splenocytes stimulated with pokeweed mitogen or concanavalin A. In anti-CD3ε-activated spleen cells, CD30 is expressed primarily on the surface of CD8+ T cells with peak expression on days 4 and 5. Stimulation of CD30+ CTL lines with plate-bound anti-CD30 directly signals IL-5 but not IFN-γ production. While these studies demonstrate that CD30 directs cytokine secretion and suggest that CD30 may play a pivotal role in the pattern of cytokine production by T cells, the precise roles of CD30 and its ligand (CD153) in T-cell development have not been clearly defined.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: mCD30.1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty