Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD38 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 90]

Supplier: Southern Biotechnology
1640-09L 1640-09
100243-470EA 371.38 USD InStock
100243-470 100243-468
Anti-CD38 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 90]
Antibodies
Murine CD38, a type II transmembrane glycoprotein, is a bifunctional ectoenzyme capable of catabolizing nicotinamide adenine dinucleotide (NAD+) to cyclic ADP-ribose (cADPR), and then hydrolyzing cADPR to adenosine diphosphoribose (ADPR). It is expressed at high levels on the surface of peripheral B-lineage cells and at low density on germinal center B cells from unimmunized mice. It has also been reported to be expressed at moderate levels on NK cells, a proportion of peripheral T cells, and a subpopulation of thymocytes which are mostly TCRαβ+, CD4-, CD8-. Murine CD38 is also expressed by all Mac-1+ macrophages in the peritoneal cavities of unimmunized mice but not by unstimulated bone-marrow-derived macrophages. Monoclonal antibodies to CD38 have been shown to induce B and T cell proliferation, protect B cells from apoptosis, and inhibit B lymphopoiesis.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 90
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty