Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SJ25-C1]

Supplier: Southern Biotechnology
9340-02 9340-02S
100245-892EA 149.08 USD
100245-892 103265-696
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SJ25-C1]
Antibodies
CD19 is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily. It is expressed at all stages of B cell differentiation except terminally differentiated plasma cells. It is also present on the cell surface of follicular dendritic cells. CD19 associates with CD21, CD81, Leu 13 and/or MHC Class II molecules to form a signal transduction complex on the B cell surface. This signaling complex modulates the activation threshold for the B cell antigen receptor (BCR).

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25-C1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR