Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHT1]

Supplier: Southern Biotechnology
9515-02 9515-02S
100246-030EA 141.23 USD InStock
100246-030 103260-170
Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHT1]
Antibodies
CD3, a member of the immunoglobulin superfamily of cell surface receptors, is comprised of five invariable chains ranging from 16-28 kDa and is closely associated with the T cell antigen receptor (TCR). CD3 is expressed on 70-80% of normal peripheral blood lymphocytes and on 10-20% of thymocytes. It plays a major role in signaling during antigen recognition, leading to T cell activation. The monoclonal antibody UCHT1 reacts with the 20 kDa ε chain of the CD3/TCR complex.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty