Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: RFT4]

Supplier: Southern Biotechnology
9522-09 9522-09S
100246-054EA 185.69 USD InStock
100246-054 103260-186
Anti-CD4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: RFT4]
Antibodies
CD4 is a 55 kDa type I transmembrane glycoprotein and a member of the Ig-SF of cell surface receptors. It is expressed predominantly on the “helper/inducer” subpopulation of mature T lymphocytes and on monocytes and macrophages. Domains 1 and 2 bind to MHC Class II antigens, while domains 3 and 4 may be involved in cis interactions with the CD3/TCR complex. CD4 functions as an accessory molecule in the recognition of foreign antigens in association with MHC class II antigens by T cells.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RFT4
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty