Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHT2]

Supplier: Southern Biotechnology
9526-02 9526-02S
100246-070EA 239.31 USD InStock
100246-070 103260-196
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHT2]
Antibodies
CD5, a member of the scavenger receptor superfamily of cell surface receptors, is a type I transmembrane glycoprotein whose ligand is CD72. It is expressed on most thymocytes, all mature T cells, and a subset of B cells. CD5 plays a role in regulation of T cell activation and signaling.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus, Cynomolgus, Chimpanzee

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty