Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD95 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DX2]

Supplier: Southern Biotechnology
9730-09 9730-09S
100246-394EA 371.38 USD InStock
100246-394 103260-222
Anti-CD95 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: DX2]
Antibodies
CD95, also known as Fas and APO-1, is a 45 kDa type I transmembrane glycoprotein and a member of the tumor necrosis factor receptor superfamily. It is expressed by activated lymphocytes, monocytes, neutrophils, fibroblasts, and cell lines. Fas ligand binding to CD95 induces apoptosis in activated mature lymphocytes thereby playing a role in maintaining peripheral tolerance. Crosslinking of CD95 by the monoclonal antibodies DX2 and DX3 delivers an apoptotic signal to Fas-sensitive cells indicating that these monoclonal antibodies recognize a functional epitope of CD95.

Type: Primary
Antigen: CD95
Clonality: Monoclonal
Clone: DX2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rhesus, African Green Monkey, Sooty Mangabey

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty