Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD13 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 22A5]

Supplier: Southern Biotechnology
9556-09 9556-09S
101442-956EA 299.46 USD InStock
101442-956 103265-318
Anti-CD13 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 22A5]
Antibodies
CD13, also known as aminopeptidase N, is a 150 kDa type II integral membrane glycoprotein. It is expressed on granulocytes, monocytes and their precursors, and on mast cells. CD13 is a zinc-binding metalloprotease that is involved in cell surface antigen presentation via its digestion of many MHC-bound regulatory peptides.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: 22A5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty