Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RFT8]

Supplier: Southern Biotechnology
9536-11 9536-11S
100246-110EA 363.54 USD InStock
100246-110 103265-280
Anti-CD8 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RFT8]
Antibodies
CD8 is a 33 kDa transmembrane glycoprotein expressed as either a CD8αα homodimer or CD8αβ heterodimer. It is expressed on the “cytotoxic/suppressor” subpopulation of peripheral T cells. CD8 functions primarily as a coreceptor with MHC class I-restricted TCR’s in antigen recognition.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: RFT8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty