Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ZAP-70 Mouse Monoclonal Antibody [clone: SBZAP]

Supplier: Southern Biotechnology
10650-01L 10650-01S
100242-614EA 235.38 USD InStock
100242-614 100242-616
Anti-ZAP-70 Mouse Monoclonal Antibody [clone: SBZAP]
Antibodies
ZAP-70 is a protein tyrosine kinase (PTK) with the primary role of catalyzing the phosphorylation of tyrosine residues in cellular proteins, and thus, contributes significantly to the initiation of cell signaling. ZAP-70 is normally expressed on T cell and NK cells.

Type: Primary
Antigen: ZAP-70
Clonality: Monoclonal
Clone: SBZAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty