Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG (H+L) Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
1031-09 1031-09S
100242-510EA 227.54 USD InStock
100242-510 100242-512
Anti-IgG (H+L) Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Mouse IgG(H+L)-PE antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty