Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG2a Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB84a]

Supplier: Southern Biotechnology
1155-09L 1155-09
103259-842EA 438.08 USD InStock
103259-842 103264-510
Anti-IgG2a Rat Antibody (RPE (R-Phycoerythrin)) [clone: SB84a]
Antibodies
PE Anti-Mouse IgG2a antibody for use in flow cytometry assays.

Type: Secondary
Antigen: IgG2a
Clonality: Monoclonal
Clone: SB84a
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty