Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CID-2858522 ≥98%

Supplier: CAYMAN CHEMICAL COMPANY, INC.
Danger

14122-1 14122-5 14122-10 14122-25
76292-998EA 45.88 USD InStock
76292-998 76293-000 76293-002 76293-004
CID-2858522 ≥98%
CID-2858522
  • A selective inhibitor of NF-κB activation

A selective inhibitor of PKC-induced NF-κB pathway activation (IC50 = 0.07 μM); inhibits IL-8 production in HEK293 cells (IC50 ≤ 0.1 μM), attenuates production of IL-2 by Jurkat T cells (IC50 ≤ 5 μM), and prevents proliferation of mouse B-lymphocytes (IC50 = ∼2 μM)


Formula: C₂₈H₃₉N₃O₃
MW: 465.6 g/mol
Storage Temperature: Freezer
CAS Number: 758679-97-9
UN: 2811
ADR: 6.1 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b6230a50d997780c351c4c7ee64c36328b2c8e2&vid=9b6230a50d997780c351c4c7ee64c36328b2c8e2&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=611abb994e0267a7b1b5138b2d829c28db933c2c&vid=611abb994e0267a7b1b5138b2d829c28db933c2c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Formulation A crystalline solid
Purity ≥98%
Stability ≥ 1 year

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty