Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NF-κB Activation Inhibitor III ≥99%

Supplier: CAYMAN CHEMICAL COMPANY, INC.

19083-5 19083-25
76293-014EA 106.81 USD InStock
76293-014 76293-016
NF-κB Activation Inhibitor III ≥99%
NF-κB Activation Inhibitor III
  • Inhibitor of TNF-α-induced MMP-9 protein expression

Inhibits TNF-α-induced MMP-9 protein expression (maximal effect at 10 µM) by blocking NF-κB activity in HT1080 human fibrosarcoma cells


Formula: C₁₀H₅ClN₄O₅S
MW: 328.6885 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00834007
CAS Number: 380623-76-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=708cfc24a020d616369104cfa8ce1b9a2c04bd26&vid=708cfc24a020d616369104cfa8ce1b9a2c04bd26&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d57ab84dd3851ceb362c170bc30fc294271feff&vid=2d57ab84dd3851ceb362c170bc30fc294271feff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Formulation A white to off-white solid
Purity ≥99%
Stability ≥ 1 year

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty