Echinocandin B
Supplier: CAYMAN CHEMICAL COMPANY, INC.
25520-1 25520-5
76308-456EA 269.34 USD InStock
76308-456 76308-458
Echinocandin B
Proteins and Peptides
Echinocandin B inhibits 1,3-β glucan synthase; inhibits incorporation of radiolabeled glucose into alkali-insoluble glucan by S. cerevisiae spheroplasts; inhibits growth of C. albicans in vitro (MIC = 3.9 µg/ml after 2 days)
  • An echinocandin antifungal
  • Stability: ≥2 years
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty