Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Fluorophenyl)naphtho[2,3-d]oxazole-4,9-dione 95%

Supplier: AMBEED, INC
Warning

A280375-10MG A280375-1MG A280375-25MG A280375-50MG A280375-5MG
77141-150EA 63.86 USD InStock
77141-150 77141-152 77141-154 77141-156 77141-158
2-(4-Fluorophenyl)naphtho[2,3-d]oxazole-4,9-dione 95%
2-(4-Fluorophenyl)naphtho[2,3-d]oxazole-4,9-dione
Precaución: for Research Use Only


Formula: C₁₇H₈FNO₃
MDL Number: MFCD01033540
CAS Number: 192718-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty