Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D2284-1G D2284-5G
TCD2284-001GEA 51 USD InStock
TCD2284-001G TCD2284-005G
2,3-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
2,3-Dimethoxyaniline
CAS Number: 6299-67-8
MDL Number: MFCD00180769
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 93
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Formula: C₈H₁₁NO₂
MDL Number: MFCD00180769
CAS Number: 6299-67-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=030ff001657573fed102ec02b8f566084a1b0c0e&vid=030ff001657573fed102ec02b8f566084a1b0c0e&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty